SY3H3D6827DE Synthonix Corporation

5-(Trifluoromethyl)indoline

  • MDL number No MDL found
  • Cas Number : 162100-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51466.html

Safety Information

MSDS https://synthonix.com/51466.html
Availability details for
Quantity
  • Availability
  • Add to Cart