SY3H3D6827E0 Synthonix Corporation

1,3,5-Tribromo-2,4,6-trimethylbenzene

  • MDL number MFCD00155003
  • Cas Number : 608-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51668.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51668.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart