SY3H3D6827E1 Synthonix Corporation

6-(Trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyridine-2-carboxylic acid

  • Cas Number : 1774893-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51687.html

Safety Information

MSDS https://synthonix.com/51687.html
Availability details for
Quantity
  • Availability
  • Add to Cart