SY3H3D6827E2 Synthonix Corporation

(2-(1H-1,2,4-Triazol-1-yl)phenyl)methanol

  • Cas Number : 914349-48-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51719.html

Safety Information

MSDS https://synthonix.com/51719.html
Availability details for
Quantity
  • Availability
  • Add to Cart