SY3H3D6827E7 Synthonix Corporation

o-Tolyloxyacetonitrile

  • MDL number No MDL found
  • Cas Number : 50635-21-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51755.html

Safety Information

MSDS https://synthonix.com/51755.html
Availability details for
Quantity
  • Availability
  • Add to Cart