SY3H3D6827E8 Synthonix Corporation

2-(3-(Trifluoromethoxy)phenyl)ethanol

  • Cas Number : 642444-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/51953.html

Safety Information

MSDS https://synthonix.com/51953.html
Availability details for
Quantity
  • Availability
  • Add to Cart