SY3H3D6827FA Synthonix Corporation

(R)-1-(p-Tolyl)ethanamine hydrochloride

  • MDL number No MDL found
  • Cas Number : 404336-49-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52161.html

Safety Information

MSDS https://synthonix.com/52161.html
Availability details for
Quantity
  • Availability
  • Add to Cart