SY3H3D682800 Synthonix Corporation

3-(m-Tolyl)-1H-pyrazole

  • MDL number No MDL found
  • Cas Number : 149739-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52276.html

Safety Information

MSDS https://synthonix.com/52276.html
Availability details for
Quantity
  • Availability
  • Add to Cart