SY3H3D682801 Synthonix Corporation

1,2,3,4-Tetrahydroquinoline-6-carbonitrile

  • MDL number No MDL found
  • Cas Number : 50741-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/52285.html

Safety Information

MSDS https://synthonix.com/52285.html
Availability details for
Quantity
  • Availability
  • Add to Cart