SY3H3D682803 Synthonix Corporation

2-((Thiazol-2-ylamino)methyl)phenol

  • Cas Number : 13159-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52338.html

Safety Information

MSDS https://synthonix.com/52338.html
Availability details for
Quantity
  • Availability
  • Add to Cart