SY3H3D68280A Synthonix Corporation

3a,4,7,7a-Tetrahydro-4,7-ethanoisobenzofuran-1,3-dione

  • MDL number No MDL found
  • Cas Number : 6708-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52464.html

Safety Information

MSDS https://synthonix.com/52464.html
Availability details for
Quantity
  • Availability
  • Add to Cart