SY3H3D68280C Synthonix Corporation

2,4,6-Trimethylbenzonitrile

  • Cas Number : 2571-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/52505.html

Safety Information

MSDS https://synthonix.com/52505.html
Availability details for
Quantity
  • Availability
  • Add to Cart