SY3H3D68280F Synthonix Corporation

5,6,7,8-Tetrahydroquinoline-6-carboxylic acid

  • Cas Number : 1256822-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52577.html

Safety Information

MSDS https://synthonix.com/52577.html
Availability details for
Quantity
  • Availability
  • Add to Cart