SY3H3D682817 Synthonix Corporation

1-[3-(Trifluoromethyl)phenyl]propylamine

  • MDL number MFCD07784289
  • Cas Number : 473732-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Colorless to light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52754.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52754.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart