SY3H3D682866 Synthonix Corporation

1,3,5-Tribromoadamantane

  • MDL number No MDL found
  • Cas Number : 707-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/52873.html

Safety Information

MSDS https://synthonix.com/52873.html
Availability details for
Quantity
  • Availability
  • Add to Cart