SY3H3D68286A Synthonix Corporation

(1,2,3,4-Tetrahydroisoquinolin-3-yl)methanol

  • MDL number No MDL found
  • Cas Number : 63006-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/52934.html

Safety Information

MSDS https://synthonix.com/52934.html
Availability details for
Quantity
  • Availability
  • Add to Cart