SY3H3D682871 Synthonix Corporation

4-[(Trimethylsilyl)oxy]benzaldehyde

  • MDL number No MDL found
  • Cas Number : 1012-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/53091.html

Safety Information

MSDS https://synthonix.com/53091.html
Availability details for
Quantity
  • Availability
  • Add to Cart