SY3H3D682874 Synthonix Corporation

4,4,5,5-Tetramethyl-2-(selenophen-2-yl)-1,3,2-dioxaborolane

  • Cas Number : 1338368-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53170.html

Safety Information

MSDS https://synthonix.com/53170.html
Availability details for
Quantity
  • Availability
  • Add to Cart