SY3H3D682882 Synthonix Corporation

4,4,4-Trifluoro-2-(4-fluorophenyl)-3-oxobutanenitrile

  • MDL number No MDL found
  • Cas Number : 3108-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53413.html

Safety Information

MSDS https://synthonix.com/53413.html
Availability details for
Quantity
  • Availability
  • Add to Cart