SY3H3D682889 Synthonix Corporation

5-(Trifluoromethyl)quinolin-8-amine

  • MDL number No MDL found
  • Cas Number : 483-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53522.html

Safety Information

MSDS https://synthonix.com/53522.html
Availability details for
Quantity
  • Availability
  • Add to Cart