SY3H3D68288A Synthonix Corporation

4-(4-(Trifluoromethyl)phenyl)oxazol-2-amine

  • MDL number No MDL found
  • Cas Number : 859721-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/53539.html

Safety Information

MSDS https://synthonix.com/53539.html
Availability details for
Quantity
  • Availability
  • Add to Cart