SY3H3D68288B Synthonix Corporation

Thiazolo[4,5-h]isoquinolin-2-amine

  • MDL number No MDL found
  • Cas Number : 35317-80-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53549.html

Safety Information

MSDS https://synthonix.com/53549.html
Availability details for
Quantity
  • Availability
  • Add to Cart