SY3H3D68288D Synthonix Corporation

2-(Thiazol-2-yl)benzaldehyde

  • MDL number No MDL found
  • Cas Number : 223575-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53590.html

Safety Information

MSDS https://synthonix.com/53590.html
Availability details for
Quantity
  • Availability
  • Add to Cart