SY3H3D682893 Synthonix Corporation

4-(Trifluoromethoxy)cinnamic Acid

  • MDL number No MDL found
  • Cas Number : 783-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53609.html

Safety Information

MSDS https://synthonix.com/53609.html
Availability details for
Quantity
  • Availability
  • Add to Cart