SY3H3D682896 Synthonix Corporation

1,1,1-Trifluoro-4-phenylbut-3-en-2-one

  • MDL number No MDL found
  • Cas Number : 86571-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53624.html

Safety Information

MSDS https://synthonix.com/53624.html
Availability details for
Quantity
  • Availability
  • Add to Cart