SY3H3D68289A Synthonix Corporation

3-(3-((Trifluoromethyl)thio)phenyl)-1H-pyrazol-5-amine

  • Cas Number : 1912400-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53689.html

Safety Information

MSDS https://synthonix.com/53689.html
Availability details for
Quantity
  • Availability
  • Add to Cart