SY3H3D68289C Synthonix Corporation

1-(4-(Trifluoromethoxy)phenyl)propan-2-one

  • Cas Number : 1249274-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54010.html

Safety Information

MSDS https://synthonix.com/54010.html
Availability details for
Quantity
  • Availability
  • Add to Cart