SY3H3D68289F Synthonix Corporation

3-(3-(Trifluoromethyl)phenyl)propanal

  • MDL number No MDL found
  • Cas Number : 21172-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/54062.html

Safety Information

MSDS https://synthonix.com/54062.html
Availability details for
Quantity
  • Availability
  • Add to Cart