SY3H3D6828A3 Synthonix Corporation

4,5,6,7-Tetrahydrobenzo[b]thiophen-2-amine oxalate

  • MDL number No MDL found
  • Cas Number : 1216924-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54076.html

Safety Information

MSDS https://synthonix.com/54076.html
Availability details for
Quantity
  • Availability
  • Add to Cart