SY3H3D6828A7 Synthonix Corporation

1-(3-(Trifluoromethyl)phenyl)cyclopropanamine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1108698-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/54092.html

Safety Information

MSDS https://synthonix.com/54092.html
Availability details for
Quantity
  • Availability
  • Add to Cart