SY3H3D6828A8 Synthonix Corporation

4-(Trifluoromethyl)-2,3-dihydro-1H-inden-1-amine hydrochloride

  • Cas Number : 68755-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54093.html

Safety Information

MSDS https://synthonix.com/54093.html
Availability details for
Quantity
  • Availability
  • Add to Cart