SY3H3D6828A9 Synthonix Corporation

6-(Trifluoromethyl)-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • MDL number No MDL found
  • Cas Number : 215798-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54094.html

Safety Information

MSDS https://synthonix.com/54094.html
Availability details for
Quantity
  • Availability
  • Add to Cart