SY3H3D6828AB Synthonix Corporation

8-(Trifluoromethyl)chroman-4-amine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1392212-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54098.html

Safety Information

MSDS https://synthonix.com/54098.html
Availability details for
Quantity
  • Availability
  • Add to Cart