SY3H3D6828AC Synthonix Corporation

3-[3-(Trifluoromethyl)phenoxy]-azetidine hydrochloride

  • Cas Number : 1188374-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54099.html

Safety Information

MSDS https://synthonix.com/54099.html
Availability details for
Quantity
  • Availability
  • Add to Cart