SY3H3D6828AF Synthonix Corporation

1,2,3,4-Tetrahydroisoquinoline-1-carboxylic acid hydrochloride

  • MDL number No MDL found
  • Cas Number : 92932-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54111.html

Safety Information

MSDS https://synthonix.com/54111.html
Availability details for
Quantity
  • Availability
  • Add to Cart