SY3H3D6829BD Synthonix Corporation

2,2,4-Trimethyl-1,2,3,4-tetrahydroquinoline hydrochloride

  • MDL number No MDL found
  • Cas Number : 4071-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56618.html

Safety Information

MSDS https://synthonix.com/56618.html
Availability details for
Quantity
  • Availability
  • Add to Cart