SY3H3D6829BF Synthonix Corporation

4-Octyl Itaconate

  • MDL number No MDL found
  • Cas Number : 3133-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/57236.html

Safety Information

MSDS https://synthonix.com/57236.html
Availability details for
Quantity
  • Availability
  • Add to Cart