SY3H3D6829C4 Synthonix Corporation

(R)-1-(p-Tolyl)propan-1-amine hydrochloride

  • MDL number No MDL found
  • Cas Number : 239105-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54185.html

Safety Information

MSDS https://synthonix.com/54185.html
Availability details for
Quantity
  • Availability
  • Add to Cart