SY3H3D6829C7 Synthonix Corporation

1,2,6,7-Tetrahydro-8H-indeno[5,4-b]furan-8-one

  • MDL number No MDL found
  • Cas Number : 196597-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/54348.html

Safety Information

MSDS https://synthonix.com/54348.html
Availability details for
Quantity
  • Availability
  • Add to Cart