SY3H3D6829CB Synthonix Corporation

Trimethyl pyridine-2,4,6-tricarboxylate

  • Cas Number : 25309-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54447.html

Safety Information

MSDS https://synthonix.com/54447.html
Availability details for
Quantity
  • Availability
  • Add to Cart