SY3H3D6829CE Synthonix Corporation

1,2,3,4-Tetrahydrocyclopenta[b]indol-2-amine

  • MDL number No MDL found
  • Cas Number : 1263284-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/54483.html

Safety Information

MSDS https://synthonix.com/54483.html
Availability details for
Quantity
  • Availability
  • Add to Cart