SY3H3D6829D6 Synthonix Corporation

5,6,7,8-Tetrahydronaphthalene-1-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 4242-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/54564.html

Safety Information

MSDS https://synthonix.com/54564.html
Availability details for
Quantity
  • Availability
  • Add to Cart