SY3H3D6829D7 Synthonix Corporation

2,3,4,5-Tetrahydro-1H-benzo[4,5]imidazo[1,2-d][1,4]diazepine

  • MDL number No MDL found
  • Cas Number : 135875-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54620.html

Safety Information

MSDS https://synthonix.com/54620.html
Availability details for
Quantity
  • Availability
  • Add to Cart