SY3H3D6829D8 Synthonix Corporation

1-(1,2,3,4-Tetrahydroquinolin-6-yl)ethanone

  • MDL number No MDL found
  • Cas Number : 113961-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54634.html

Safety Information

MSDS https://synthonix.com/54634.html
Availability details for
Quantity
  • Availability
  • Add to Cart