SY3H3D6829DA Synthonix Corporation

3,5,7-Trifluoroadamantane-1-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 214557-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54679.html

Safety Information

MSDS https://synthonix.com/54679.html
Availability details for
Quantity
  • Availability
  • Add to Cart