SY3H3D6829DC Synthonix Corporation

2-(2-(2,2,2-Trifluoroethoxy)phenoxy)ethyl methanesulfonate

  • MDL number No MDL found
  • Cas Number : 160969-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/54687.html

Safety Information

MSDS https://synthonix.com/54687.html
Availability details for
Quantity
  • Availability
  • Add to Cart