SY3H3D6829E0 Synthonix Corporation

(5,6,7,8-Tetrahydronaphthalen-1-yl)methanol

  • MDL number No MDL found
  • Cas Number : 41790-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54761.html

Safety Information

MSDS https://synthonix.com/54761.html
Availability details for
Quantity
  • Availability
  • Add to Cart