SY3H3D6829E3 Synthonix Corporation

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)pyrimidin-2-amine

  • MDL number No MDL found
  • Cas Number : 944401-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/54841.html

Safety Information

MSDS https://synthonix.com/54841.html
Availability details for
Quantity
  • Availability
  • Add to Cart