SY3H3D6829E4 Synthonix Corporation

4-(Tetrahydropyran-4-yl)phenylamine

  • MDL number No MDL found
  • Cas Number : 62071-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54858.html

Safety Information

MSDS https://synthonix.com/54858.html
Availability details for
Quantity
  • Availability
  • Add to Cart