SY3H3D6829EC Synthonix Corporation

6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2(1H)-one

  • MDL number No MDL found
  • Cas Number : 1310405-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54946.html

Safety Information

MSDS https://synthonix.com/54946.html
Availability details for
Quantity
  • Availability
  • Add to Cart